ENA429361607 Enamine

tert-butyl 3-cyanopyrrolidine-1-carboxylate

  • MDL number MFCD03001709
  • Cas Number : 476493-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68721

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-68721
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart