ENA429361612 Enamine

1-(difluoromethoxy)-2,5-difluoro-4-nitrobenzene

  • MDL number MFCD28118883
  • Cas Number : 1803610-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71516
Availability details for
Quantity
  • Availability
  • Add to Cart