ENA429361613 Enamine

methyl 2-(4-phenylcyclohexylidene)acetate

  • MDL number MFCD18374020
  • Cas Number : 920021-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73022
Availability details for
Quantity
  • Availability
  • Add to Cart