ENA429361615 Enamine

4-chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09744140
  • Cas Number : 1057672-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74721
Availability details for
Quantity
  • Availability
  • Add to Cart