ENA429361616 Enamine

1-methyl-3-(thiophen-2-yl)-1H-1,2,4-triazol-5-amine

  • MDL number MFCD16859188
  • Cas Number : 1342010-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75408

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75408
Availability details for
Quantity
  • Availability
  • Add to Cart