ENA429361617 Enamine

1-(2,3-dihydro-1H-isoindol-2-yl)ethan-1-one

  • MDL number MFCD18809201
  • Cas Number : 18913-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75418
Availability details for
Quantity
  • Availability
  • Add to Cart