ENA429361619 Enamine

3-amino-1lambda6-benzothiophene-1,1-dione

  • MDL number MFCD20547710
  • Cas Number : 858819-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76424
Availability details for
Quantity
  • Availability
  • Add to Cart