ENA429361626 Enamine

2-({[(tert-butoxy)carbonyl]amino}methyl)-3-methylbutanoic acid

  • MDL number MFCD11973915
  • Cas Number : 1233517-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78242
Availability details for
Quantity
  • Availability
  • Add to Cart