ENA429361636 Enamine

2-[cyclopropyl({[(9H-fluoren-9-yl)methoxy]carbonyl})amino]acetic acid

  • MDL number MFCD18328445
  • Cas Number : 1342767-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81167
Availability details for
Quantity
  • Availability
  • Add to Cart