ENA429361639 Enamine

2-(1-aminocyclohexyl)acetic acid

  • MDL number MFCD02258116
  • Cas Number : 37631-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83335
Availability details for
Quantity
  • Availability
  • Add to Cart