ENA429361647 Enamine

3-(oxan-4-yl)-3-oxopropanenitrile

  • MDL number MFCD17016045
  • Cas Number : 1010798-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85186

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85186
Availability details for
Quantity
  • Availability
  • Add to Cart