ENA429361652 Enamine

4-bromo-1-methyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD11640404
  • Cas Number : 83255-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253-254
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86412
Availability details for
Quantity
  • Availability
  • Add to Cart