ENA429361658 Enamine

2-[(trimethylsilyl)oxy]propanenitrile

  • MDL number MFCD16786577
  • Cas Number : 41309-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91915

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91915
Availability details for
Quantity
  • Availability
  • Add to Cart