ENA429361685 Enamine

2-methoxy-2-methylpropanenitrile

  • MDL number MFCD21648802
  • Cas Number : 76474-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-97557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97557
Availability details for
Quantity
  • Availability
  • Add to Cart