ENA429361691 Enamine

3-(5-cyclopropyl-1,2,4-oxadiazol-3-yl)aniline hydrochloride

  • MDL number MFCD28118887
  • Cas Number : 1803603-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99669
Availability details for
Quantity
  • Availability
  • Add to Cart