ENA432537866 Enamine

1-methyl-3-(piperidin-3-yl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD28145194
  • Cas Number : 1803593-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-171833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171833
Availability details for
Quantity
  • Availability
  • Add to Cart