ENA432537870 Enamine

(1R)-1-(4-methylpyrimidin-5-yl)ethan-1-amine

  • MDL number MFCD28139361
  • Cas Number : 1704940-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177593
Availability details for
Quantity
  • Availability
  • Add to Cart