ENA432537881 Enamine

3-bromo-4-methyl-2-nitroaniline

  • MDL number MFCD28139367
  • Cas Number : 1803582-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183187
Availability details for
Quantity
  • Availability
  • Add to Cart