ENA432537886 Enamine

3,3-dimethyl-1-(2-methyl-5-nitrophenyl)urea

  • MDL number MFCD28139373
  • Cas Number : 1696207-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185882
Availability details for
Quantity
  • Availability
  • Add to Cart