ENA432537887 Enamine

5-bromo-1-[(5-ethyl-1,2,4-oxadiazol-3-yl)methyl]-2-methyl-1,4-dihydropyrimidin-4-one

  • MDL number MFCD28145216
  • Cas Number : 1803609-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187318
Availability details for
Quantity
  • Availability
  • Add to Cart