ENA432537889 Enamine

2-[(benzyloxy)methyl]-1,3,4-oxadiazole

  • MDL number MFCD28145218
  • Cas Number : 23571-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187926
Availability details for
Quantity
  • Availability
  • Add to Cart