ENA432537890 Enamine

2-(2-methoxyethyl)-1,3-dithiane

  • MDL number MFCD28145221
  • Cas Number : 938184-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188545
Availability details for
Quantity
  • Availability
  • Add to Cart