ENA432537891 Enamine

3-chloro-5-(chlorosulfonyl)-2-methylbenzoic acid

  • MDL number MFCD28145222
  • Cas Number : 1702890-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188566
Availability details for
Quantity
  • Availability
  • Add to Cart