ENA432537897 Enamine

5-cyanothiophene-2-sulfonyl chloride

  • MDL number MFCD28145224
  • Cas Number : 1803598-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189705
Availability details for
Quantity
  • Availability
  • Add to Cart