ENA432537898 Enamine

2-(2,3-dihydro-1-benzofuran-3-yl)ethanimidamide hydrochloride

  • MDL number MFCD28139380
  • Cas Number : 1803600-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189790
Availability details for
Quantity
  • Availability
  • Add to Cart