ENA432537899 Enamine

1-[(2,2-dimethylpropyl)sulfanyl]ethan-1-one

  • MDL number MFCD28145225
  • Cas Number : 1803600-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189853
Availability details for
Quantity
  • Availability
  • Add to Cart