ENA432537900 Enamine

4-(3-chloro-2-oxopropyl)phenyl acetate

  • MDL number MFCD28139381
  • Cas Number : 1803594-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189915
Availability details for
Quantity
  • Availability
  • Add to Cart