ENA432537904 Enamine

1-(benzyloxy)-2-chloro-4-(difluoromethyl)benzene

  • MDL number MFCD28145228
  • Cas Number : 1803586-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190642
Availability details for
Quantity
  • Availability
  • Add to Cart