ENA432537910 Enamine

ethyl 3-cyclohexyl-2-fluoropropanoate

  • MDL number MFCD28139390
  • Cas Number : 1803587-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191393
Availability details for
Quantity
  • Availability
  • Add to Cart