ENA432537913 Enamine

2-{[(octahydro-1H-quinolizin-1-yl)methyl]sulfanyl}acetic acid hydrochloride

  • MDL number MFCD28139393
  • Cas Number : 1803611-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191517
Availability details for
Quantity
  • Availability
  • Add to Cart