ENA432537919 Enamine

1,2-dimethyl-5-(methylsulfanyl)-3-nitrobenzene

  • MDL number MFCD28139399
  • Cas Number : 1803607-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191618
Availability details for
Quantity
  • Availability
  • Add to Cart