ENA432537922 Enamine

ethyl 3-({[(benzyloxy)carbonyl]amino}methyl)-1H-1,2,4-triazole-5-carboxylate

  • MDL number MFCD28139402
  • Cas Number : 1803566-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191750
Availability details for
Quantity
  • Availability
  • Add to Cart