ENA432537923 Enamine

2-[(5-methyl-1,3-oxazol-2-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD28139403
  • Cas Number : 1133256-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191775
Availability details for
Quantity
  • Availability
  • Add to Cart