ENA432537924 Enamine

1-benzyl-3-methylpyrrolidine-3-carboxamide

  • MDL number MFCD28139404
  • Cas Number : 1803588-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191834
Availability details for
Quantity
  • Availability
  • Add to Cart