ENA432537926 Enamine

3-(bromomethyl)pyridin-2-amine hydrobromide

  • MDL number MFCD28145231
  • Cas Number : 23612-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191965
Availability details for
Quantity
  • Availability
  • Add to Cart