ENA432537936 Enamine

(1R)-1-(5-methyl-1,2-oxazol-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD28139417
  • Cas Number : 1807921-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192795
Availability details for
Quantity
  • Availability
  • Add to Cart