ENA432537937 Enamine

(1R)-1-(5-methyl-1,2-oxazol-3-yl)ethan-1-ol

  • MDL number MFCD28139418
  • Cas Number : 1807938-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192796
Availability details for
Quantity
  • Availability
  • Add to Cart