ENA432537938 Enamine

(2R,3R)-1-methyl-2-(1-methyl-1H-pyrazol-4-yl)-5-oxopyrrolidine-3-carboxamide

  • MDL number MFCD28657005
  • Cas Number : 1820569-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192826
Availability details for
Quantity
  • Availability
  • Add to Cart