ENA432537939 Enamine

4-(1-methyl-1H-pyrazol-4-yl)-4-oxobut-2-enoic acid

  • Cas Number : 1807920-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192851
Availability details for
Quantity
  • Availability
  • Add to Cart