ENA432537942 Enamine

(1S)-1-(4-methylpyrimidin-5-yl)ethan-1-ol

  • MDL number MFCD28139423
  • Cas Number : 1704959-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192889
Availability details for
Quantity
  • Availability
  • Add to Cart