ENA432537945 Enamine

3-ethyl-3-methyl-2,3,4,5-tetrahydro-1H-1,4-benzodiazepin-5-one

  • MDL number MFCD28139426
  • Cas Number : 1803571-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192916
Availability details for
Quantity
  • Availability
  • Add to Cart