ENA432537946 Enamine

3-ethyl-3-methyl-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine dihydrochloride

  • MDL number MFCD28139427
  • Cas Number : 1803587-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192918
Availability details for
Quantity
  • Availability
  • Add to Cart