ENA432537950 Enamine

(2S)-2-amino-1-(2,3-dihydro-1H-isoindol-2-yl)propan-1-one hydrochloride

  • MDL number MFCD28139434
  • Cas Number : 1690144-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193007
Availability details for
Quantity
  • Availability
  • Add to Cart