ENA432537951 Enamine

2-amino-1-(2,3-dihydro-1H-isoindol-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD28139435
  • Cas Number : 1803591-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193009
Availability details for
Quantity
  • Availability
  • Add to Cart