ENA432537952 Enamine

methyl (3aR,7aS)-2-benzyl-5-methyl-octahydro-1H-pyrrolo[3,4-c]pyridine-7a-carboxylate

  • MDL number MFCD28139437
  • Cas Number : 1820570-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193017
Availability details for
Quantity
  • Availability
  • Add to Cart