ENA432537960 Enamine

2-{[(1-ethyl-1H-pyrazol-4-yl)methyl]amino}ethan-1-ol hydrochloride

  • MDL number MFCD28139449
  • Cas Number : 1803590-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193133
Availability details for
Quantity
  • Availability
  • Add to Cart