ENA432537961 Enamine

bicyclo[3.1.0]hexan-2-amine hydrochloride

  • MDL number MFCD28139450
  • Cas Number : 1803596-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193138
Availability details for
Quantity
  • Availability
  • Add to Cart