ENA432537968 Enamine

3-methyl-3-(1H-1,2,4-triazol-1-yl)butan-2-one

  • MDL number MFCD28139465
  • Cas Number : 1803587-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193242
Availability details for
Quantity
  • Availability
  • Add to Cart