ENA432537973 Enamine

4-chloro-1-(methoxymethyl)-1H-pyrazole-5-sulfonyl chloride

  • MDL number MFCD28145241
  • Cas Number : 1803611-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193361
Availability details for
Quantity
  • Availability
  • Add to Cart