ENA432537974 Enamine

1-[2-(4-chlorophenyl)ethyl]-5-nitro-1H-1,3-benzodiazole

  • MDL number MFCD28139475
  • Cas Number : 1803599-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193371
Availability details for
Quantity
  • Availability
  • Add to Cart